Assay Details
Gene Symbol: KXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194825.1 | NP_001181754.1 | 336 | 62 | ||
XM_015123314.2 | XP_014978800.1 | 435 | 62 | |||
XM_028838552.1 | XP_028694385.1 | 459 | 62 | |||
GenBank mRNA | JU336339.1 | - | 338 | 62 | ||
JU477162.1 | - | 398 | 62 | |||
JV045587.1 | - | 321 | 62 | |||
JV529003.1 | - | 360 | 62 |

More Information
Set Membership: |
![]() ![]() |
