Assay Details
Gene Symbol: FA2H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194422.3 | NP_001181351.2 | 1 - 2 | 351 | 98 | |
GenBank mRNA | JU336356.1 | - | 1 - 2 | 332 | 98 | |
JV723233.1 | - | 1 - 2 | 258 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() |
