Assay Details
Gene Symbol: TNIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137874.2 | XP_014993360.1 | 1127 | 166 | ||
XM_015137875.2 | XP_014993361.1 | 1127 | 166 | |||
XM_015137876.2 | XP_014993362.1 | 878 | 166 | |||
XM_015137877.2 | XP_014993363.1 | 878 | 166 | |||
GenBank mRNA | JU493311.1 | - | 848 | 166 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
