Assay Details
Gene Symbol: RIPOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126402.2 | XP_014981888.1 | 515 | 135 | ||
XM_015126404.2 | XP_014981890.1 | 493 | 135 | |||
XM_015126405.1 | XP_014981891.1 | 567 | 135 | |||
XM_028841362.1 | XP_028697195.1 | 940 | 135 | |||
XM_028841364.1 | XP_028697197.1 | 509 | 135 | |||
GenBank mRNA | JV557734.1 | - | 5 - 6 | 470 | 135 | |
JV628622.1 | - | 5 - 6 | 470 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
