Assay Details
Gene Symbol: MUL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261655.1 | NP_001248584.1 | 233 | 138 | ||
GenBank mRNA | JU320766.1 | - | 239 | 138 | ||
JV044957.1 | - | 233 | 138 |

More Information
Set Membership: |
![]() ![]() |
