Assay Details
Gene Symbol: OCEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194595.2 | NP_001181524.2 | 1 - 2 | 111 | 123 | |
GenBank mRNA | JV481845.1 | - | 1 - 2 | 64 | 123 | |
JV593627.1 | - | 1 - 2 | 54 | 123 | ||
JV628728.1 | - | 1 - 2 | 47 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
