Assay Details
Gene Symbol: OGFOD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261553.1 | NP_001248482.1 | 826 | 107 | ||
GenBank mRNA | JU336408.1 | - | 881 | 107 | ||
JV044917.1 | - | 826 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
