Assay Details
Gene Symbol: ALG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015115675.2 | XP_014971161.2 | 658 | 81 | ||
XM_015115678.2 | XP_014971164.1 | 658 | 81 | |||
XM_028833961.1 | XP_028689794.1 | 658 | 81 | |||
XM_028833962.1 | XP_028689795.1 | 658 | 81 | |||
XM_028833963.1 | XP_028689796.1 | 573 | 81 | |||
XM_028833964.1 | XP_028689797.1 | 651 | 81 | |||
XM_028833965.1 | XP_028689798.1 | 786 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
