Assay Details
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261154.1 | NP_001248083.1 | 1 - 2 | 257 | 139 | |
GenBank mRNA | JU336433.1 | - | 1 - 2 | 249 | 139 | |
JU477249.1 | - | 1 - 2 | 226 | 139 | ||
JV048280.1 | - | 1 - 2 | 257 | 139 | ||
JV602242.1 | - | 1 - 2 | 306 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
