Assay Details
Gene Symbol: PIP4K2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261154.1 | NP_001248083.1 | 7 - 8 | 894 | 157 | |
GenBank mRNA | JU336433.1 | - | 7 - 8 | 886 | 157 | |
JU477249.1 | - | 7 - 8 | 863 | 157 | ||
JV048280.1 | - | 7 - 8 | 894 | 157 | ||
JV602242.1 | - | 7 - 8 | 943 | 157 |

More Information
Set Membership: |
![]() ![]() ![]() |
