Assay Details
Gene Symbol: PDZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104508.4 | XP_001104508.1 | 577 | 75 | ||
XM_001104665.4 | XP_001104665.1 | 577 | 75 | |||
XM_015115951.2 | XP_014971437.1 | 546 | 75 | |||
XM_015115952.2 | XP_014971438.1 | 546 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
