Assay Details
Gene Symbol: JHY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001107883.3 | XP_001107883.3 | 2164 | 180 | ||
XM_015115978.2 | XP_014971464.2 | 2096 | 180 | |||
XM_015115979.2 | XP_014971465.2 | 1758 | 180 | |||
XM_015115982.2 | XP_014971468.2 | 1726 | 180 | |||
XM_028834095.1 | XP_028689928.1 | 1982 | 180 | |||
XM_028834096.1 | XP_028689929.1 | 2050 | 180 |

More Information
Set Membership: |
![]() ![]() |
