Assay Details
Gene Symbol: DGLUCY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015144368.2 | XP_014999854.2 | 1716 | 148 | ||
XM_015144369.2 | XP_014999855.2 | 1701 | 148 | |||
XM_015144370.2 | XP_014999856.2 | 1566 | 148 | |||
XM_015144371.2 | XP_014999857.2 | 1813 | 148 | |||
XM_015144372.2 | XP_014999858.2 | 1769 | 148 | |||
XM_028851759.1 | XP_028707592.1 | 1717 | 148 | |||
XM_028851761.1 | XP_028707594.1 | 1810 | 148 |

More Information
Set Membership: |
![]() ![]() |
