Assay Details
Gene Symbol: DBF4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015119879.2 | XP_014975365.2 | 656 | 71 | ||
XM_015119880.2 | XP_014975366.2 | 654 | 71 | |||
XM_015119881.2 | XP_014975367.2 | 665 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
