Assay Details
Gene Symbol: NAA50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261371.2 | NP_001248300.1 | 534 | 144 | ||
XM_015129813.2 | XP_014985299.1 | 683 | 144 | |||
XM_028843327.1 | XP_028699160.1 | 814 | 144 | |||
XM_028843328.1 | XP_028699161.1 | 811 | 144 | |||
GenBank mRNA | JU336491.1 | - | 637 | 144 | ||
JU471806.1 | - | 543 | 144 | |||
JV043838.1 | - | 536 | 144 | |||
JV637198.1 | - | 637 | 144 | |||
JV697188.1 | - | 639 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
