Assay Details
Gene Symbol: LMAN2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112894.2 | XP_014968380.2 | 675 | 88 | ||
XM_028832173.1 | XP_028688006.1 | 758 | 88 | |||
XM_028832174.1 | XP_028688007.1 | 725 | 88 | |||
XM_028832175.1 | XP_028688008.1 | 839 | 88 | |||
XM_028832176.1 | XP_028688009.1 | 756 | 88 | |||
XM_028832177.1 | XP_028688010.1 | 806 | 88 | |||
XM_028832178.1 | XP_028688011.1 | 642 | 88 | |||
XM_028832179.1 | XP_028688012.1 | 723 | 88 | |||
GenBank mRNA | JU328489.1 | - | 5 - 6 | 626 | 88 | |
JV521776.1 | - | 5 - 6 | 510 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
