Assay Details
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266074.1 | NP_001253003.1 | 2 - 3 | 202 | 137 | |
XM_015120353.2 | XP_014975839.2 | 624 | 137 | |||
XM_015120354.2 | XP_014975840.2 | 574 | 137 | |||
XM_015120355.2 | XP_014975841.2 | 551 | 137 | |||
XM_015120356.2 | XP_014975842.2 | 329 | 137 | |||
XM_028835576.1 | XP_028691409.1 | 522 | 137 | |||
GenBank mRNA | JU473469.1 | - | 2 - 3 | 231 | 137 | |
JV045734.1 | - | 2 - 3 | 202 | 137 | ||
JV641575.1 | - | 2 - 3 | 396 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
