Assay Details
Gene Symbol: DOHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100385.4 | XP_001100385.1 | 517 | 83 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_001100385.4 transcripts.
This assay is in the 5'UTR region of the XM_001100385.4 transcripts.
Set Membership: |
![]() ![]() |
