Assay Details
Gene Symbol: IL20RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194658.3 | NP_001181587.2 | 334 | 124 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
