Assay Details
Gene Symbol: STOML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193787.2 | NP_001180716.2 | 4 - 5 | 663 | 119 | |
GenBank mRNA | JV643738.1 | - | 2 - 3 | 141 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
