Assay Details
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091148.4 | XP_001091148.3 | 659 | 111 | ||
XM_015126826.2 | XP_014982312.1 | 637 | 111 | |||
GenBank mRNA | JU325623.1 | - | 2 - 3 | 349 | 111 | |
JV668364.1 | - | 3 - 4 | 611 | 111 |
Gene Symbol: FANCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU325622.1 | - | 2 - 3 | 349 | 111 | |
JU474667.1 | - | 50 - 51 | 6005 | 111 | ||
JU474668.1 | - | 49 - 50 | 6077 | 111 | ||
JU474670.1 | - | 50 - 51 | 6005 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
