Assay Details
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091148.4 | XP_001091148.3 | 1186 | 116 | ||
XM_015126826.2 | XP_014982312.1 | 1164 | 116 | |||
GenBank mRNA | JU325623.1 | - | 4 - 5 | 876 | 116 | |
JV668364.1 | - | 5 - 6 | 1138 | 116 |
Gene Symbol: FANCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU325622.1 | - | 4 - 5 | 876 | 116 | |
JU474667.1 | - | 48 - 49 | 5478 | 116 | ||
JU474668.1 | - | 47 - 48 | 5550 | 116 | ||
JU474670.1 | - | 48 - 49 | 5478 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() |
