Assay Details
Gene Symbol: ZNF276 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001091148.4 | XP_001091148.3 | 1679 | 85 | ||
XM_015126826.2 | XP_014982312.1 | 1657 | 85 | |||
GenBank mRNA | JU325623.1 | - | 9 - 10 | 1369 | 85 | |
JV668364.1 | - | 10 - 11 | 1631 | 85 |
Gene Symbol: FANCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU325622.1 | - | 9 - 10 | 1369 | 85 | |
JU474667.1 | - | 43 - 44 | 4985 | 85 | ||
JU474669.1 | - | 43 - 44 | 4985 | 85 | ||
JU474670.1 | - | 43 - 44 | 4985 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
