Assay Details
Gene Symbol: B3GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126896.2 | XP_014982382.2 | 779 | 73 | ||
XM_028849542.1 | XP_028705375.1 | 772 | 73 | |||
GenBank mRNA | JU326496.1 | - | 508 | 73 | ||
JU474413.1 | - | 715 | 73 | |||
JU474415.1 | - | 715 | 73 |
Gene Symbol: TBCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU474414.1 | - | 715 | 73 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
