Assay Details
Gene Symbol: B3GALNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015126896.2 | XP_014982382.2 | 1585 | 102 | ||
GenBank mRNA | JU326496.1 | - | 1314 | 102 | ||
JU474413.1 | - | 1521 | 102 | |||
JU474415.1 | - | 1521 | 102 |
Gene Symbol: TBCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU474414.1 | - | 1521 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
