Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194030.3 | NP_001180959.2 | 14 - 15 | 2186 | 138 | |
XM_028844953.1 | XP_028700786.1 | 2181 | 138 | |||
XM_028844955.1 | XP_028700788.1 | 2154 | 138 | |||
GenBank mRNA | JU321423.1 | - | 14 - 15 | 2124 | 138 | |
JV709601.1 | - | 14 - 15 | 1944 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() |
