Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194030.3 | NP_001180959.2 | 19 - 20 | 2869 | 134 | |
XM_028844953.1 | XP_028700786.1 | 2864 | 134 | |||
XM_028844954.1 | XP_028700787.1 | 2690 | 134 | |||
XM_028844955.1 | XP_028700788.1 | 2837 | 134 | |||
GenBank mRNA | JU321423.1 | - | 19 - 20 | 2807 | 134 | |
JV709601.1 | - | 19 - 20 | 2627 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
