Assay Details
Gene Symbol: ELAPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194030.3 | NP_001180959.2 | 10 - 11 | 1587 | 80 | |
XM_028844953.1 | XP_028700786.1 | 1582 | 80 | |||
XM_028844954.1 | XP_028700787.1 | 1582 | 80 | |||
XM_028844955.1 | XP_028700788.1 | 1555 | 80 | |||
GenBank mRNA | JU321423.1 | - | 10 - 11 | 1525 | 80 | |
JV709601.1 | - | 10 - 11 | 1345 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
