Assay Details
Gene Symbol: KCTD20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257899.1 | NP_001244828.1 | 1122 | 161 | ||
XM_015136134.2 | XP_014991620.1 | 823 | 161 | |||
XM_028846873.1 | XP_028702706.1 | 1209 | 161 | |||
XM_028846874.1 | XP_028702707.1 | 680 | 161 | |||
GenBank mRNA | JU333420.1 | - | 1122 | 161 | ||
JU474701.1 | - | 1049 | 161 | |||
JV046459.1 | - | 1108 | 161 |
Gene Symbol: LOC114677752 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028847910.1 | XP_028703743.1 | 161 | 161 |

More Information
Set Membership: |
![]() ![]() ![]() |
