Assay Details
Gene Symbol: RELL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193999.1 | NP_001180928.1 | 1031 | 72 | ||
XM_028849194.1 | XP_028705027.1 | 1031 | 72 | |||
GenBank mRNA | JU324667.1 | - | 1001 | 72 | ||
JV614063.1 | - | 1040 | 72 | |||
JV614064.1 | - | 1033 | 72 | |||
JV667382.1 | - | 1447 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
