Assay Details
Gene Symbol: FAAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095907.4 | XP_001095907.1 | 987 | 147 | ||
XM_015127556.2 | XP_014983042.1 | 777 | 147 | |||
XM_015127557.2 | XP_014983043.1 | 1112 | 147 | |||
GenBank mRNA | JU481280.1 | - | 2 - 3 | 245 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() |
