Assay Details
Gene Symbol: FAAH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001095907.4 | XP_001095907.1 | 1242 | 115 | ||
XM_015127556.2 | XP_014983042.1 | 1032 | 115 | |||
XM_015127557.2 | XP_014983043.1 | 1367 | 115 | |||
GenBank mRNA | JU481280.1 | - | 4 - 5 | 500 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
