Assay Details
Gene Symbol: SMIM14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015138213.2 | XP_014993699.1 | 825 | 131 | ||
XM_015138214.2 | XP_014993700.1 | 650 | 131 | |||
GenBank mRNA | JU321591.1 | - | 425 | 131 | ||
JU474628.1 | - | 417 | 131 | |||
JV046408.1 | - | 403 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
