Assay Details
Gene Symbol: ZNF598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125338.2 | XP_014980824.2 | 748 | 100 | ||
XM_015125339.2 | XP_014980825.2 | 730 | 100 | |||
XM_015125340.2 | XP_014980826.2 | 303 | 100 | |||
GenBank mRNA | JV632966.1 | - | 4 - 5 | 667 | 100 | |
JV632967.1 | - | 4 - 5 | 667 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
