Assay Details
Gene Symbol: TAFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152189.2 | XP_015007675.1 | 1589 | 89 | ||
GenBank mRNA | JU320216.1 | - | 1 - 2 | 381 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015152189.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
