Assay Details
Gene Symbol: TAFA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015152189.2 | XP_015007675.1 | 1969 | 92 | ||
XM_015152191.2 | XP_015007677.1 | 1664 | 92 | |||
XM_015152192.2 | XP_015007678.1 | 404 | 92 | |||
XM_028829685.1 | XP_028685518.1 | 802 | 92 | |||
GenBank mRNA | JU320216.1 | - | 4 - 5 | 761 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
