Assay Details
Gene Symbol: IL22RA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015137361.2 | XP_014992847.1 | 505 | 83 | ||
XM_028847154.1 | XP_028702987.1 | 505 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
