Assay Details
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111535.2 | XP_014967021.1 | 493 | 139 | ||
GenBank mRNA | JU323738.1 | - | 87 | 139 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015111535.2 transcripts.
Set Membership: |
![]() ![]() |
