Assay Details
Gene Symbol: TMPRSS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151717.2 | XP_015007203.1 | 758 | 140 | ||
XM_015151718.2 | XP_015007204.1 | 702 | 140 |

More Information
Set Membership: |
![]() ![]() |
