Assay Details
Gene Symbol: TMPRSS11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028848176.1 | XP_028704009.1 | 1227 | 155 | ||
XM_028848177.1 | XP_028704010.1 | 1218 | 155 | |||
XM_028848178.1 | XP_028704011.1 | 1024 | 155 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
