Assay Details
Gene Symbol: GLT6D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001097091.4 | XP_001097091.1 | 406 | 108 | ||
XM_001097205.4 | XP_001097205.1 | 362 | 108 | |||
XM_028834614.1 | XP_028690447.1 | 407 | 108 | |||
XM_028834615.1 | XP_028690448.1 | 596 | 108 |

More Information
Set Membership: |
![]() ![]() |
