Assay Details
Gene Symbol: LYPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194767.3 | NP_001181696.1 | 3 - 4 | 468 | 69 | |
GenBank mRNA | JV045566.1 | - | 3 - 4 | 372 | 69 | |
JV635960.1 | - | 3 - 4 | 369 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
