Assay Details
Gene Symbol: LYPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194767.3 | NP_001181696.1 | 589 | 118 | ||
GenBank mRNA | JV045566.1 | - | 493 | 118 | ||
JV635960.1 | - | 490 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
