Assay Details
Gene Symbol: RPS19BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099808.4 | XP_001099808.2 | 394 | 77 | ||
GenBank mRNA | JU477790.1 | - | 355 | 77 | ||
JV700997.1 | - | 356 | 77 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
