Assay Details
Gene Symbol: SPACA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028840153.1 | XP_028695986.1 | 474 | 96 | ||
XM_028840154.1 | XP_028695987.1 | 268 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
