Assay Details
Gene Symbol: ALKAL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194437.2 | NP_001181366.1 | 191 | 130 | ||
XM_015145342.2 | XP_015000828.1 | 1438 | 130 |

More Information
Set Membership: |
![]() ![]() |
