Assay Details
Gene Symbol: CDKN1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015113372.2 | XP_014968858.2 | 143 | 79 | ||
XM_028833066.1 | XP_028688899.1 | 143 | 79 | |||
XM_028833068.1 | XP_028688901.1 | 143 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015113372.2, XM_028833066.1, XM_028833068.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
