Assay Details
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086002.3 | XP_001086002.2 | 336 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001086002.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
