Assay Details
Gene Symbol: BIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108040.4 | XP_001108040.2 | 569 | 147 | ||
XM_015150513.2 | XP_015005999.2 | 352 | 147 | |||
XM_015150514.2 | XP_015006000.2 | 572 | 147 | |||
GenBank mRNA | JV658414.1 | - | 2 - 3 | 186 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
