Assay Details
Gene Symbol: CREBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261237.1 | NP_001248166.1 | 260 | 96 | ||
GenBank mRNA | JU320518.1 | - | 287 | 96 | ||
JU474359.1 | - | 287 | 96 | |||
JV046229.1 | - | 260 | 96 | |||
JV571724.1 | - | 299 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001261237.1 transcripts.
Set Membership: |
![]() ![]() |
